MilliporeSigma
  • Solvent-and catalysts-free immobilization of tannic acid and polyvinylpyrrolidone onto PMMA surface by DBD plasma.
Solvent-and catalysts-free immobilization of tannic acid and polyvinylpyrrolidone onto PMMA surface by DBD plasma.
Plasma Processes and Polymers (2017)
Klebert S, et al.