MilliporeSigma
  • The plasma membrane metal-ion transporter ZIP14 contributes to nontransferrin-bound iron uptake by human β-cells.
The plasma membrane metal-ion transporter ZIP14 contributes to nontransferrin-bound iron uptake by human β-cells.
American journal of physiology. Cell physiology (2016-12-03)
Richard Coffey, Mitchell D Knutson
ABSTRACT
The relationship between iron and β-cell dysfunction has long been recognized as individuals with iron overload display an increased incidence of diabetes. This link is usually attributed to the accumulation of excess iron in β-cells leading to cellular damage and impaired function. Yet, the molecular mechanism(s) by which human β-cells take up iron has not been determined. In the present study, we assessed the contribution of the metal-ion transporters ZRT/IRT-like protein 14 and 8 (ZIP14 and ZIP8) and divalent metal-ion transporter-1 (DMT1) to iron uptake by human β-cells. Iron was provided to the cells as nontransferrin-bound iron (NTBI), which appears in the plasma during iron overload and is a major contributor to tissue iron loading. We found that overexpression of ZIP14 and ZIP8, but not DMT1, resulted in increased NTBI uptake by βlox5 cells, a human β-cell line. Conversely, siRNA-mediated knockdown of ZIP14, but not ZIP8, resulted in 50% lower NTBI uptake in βlox5 cells. In primary human islets, knockdown of ZIP14 also reduced NTBI uptake by 50%. Immunofluorescence analysis of islets from human pancreatic sections localized ZIP14 and DMT1 nearly exclusively to β-cells. Studies in primary human islets suggest that ZIP14 protein levels do not vary with iron status or treatment with IL-1β. Collectively, these observations identify ZIP14 as a major contributor to NTBI uptake by β-cells and suggest differential regulation of ZIP14 in primary human islets compared with other cell types such as hepatocytes.
Sigma-Aldrich
MISSION® esiRNA, targeting human SLC39A8
Pricing and availability is not currently available.