- Validation of a method for sedatives and β-blockers determination in swine, bovine and equine kidney using liquid chromatography coupled with tandem mass spectrometry.
Validation of a method for sedatives and β-blockers determination in swine, bovine and equine kidney using liquid chromatography coupled with tandem mass spectrometry.
A method for the determination of five sedatives and 14 β-blocker residues in swine, bovine and equine kidney was validated. Samples were extracted with acetonitrile and purified using dispersive solid phase extraction (d-SPE) with Celite 545 with subsequent analysis by LC-MS/MS. A simplified protocol was applied to validate the method scope extension to include new matrices. Parameters such as recovery, trueness, linearity (r