MilliporeSigma
  • Synthesis, Crystal Structures and Antibacterial Activities of Trifluoromethylsulfonate Salts of 2-[(3-Chloropyridinium-2-yl)hydrazonomethyl]-6-methoxyphenol and its Copper(II) and Cobalt(III) Complexes.
Synthesis, Crystal Structures and Antibacterial Activities of Trifluoromethylsulfonate Salts of 2-[(3-Chloropyridinium-2-yl)hydrazonomethyl]-6-methoxyphenol and its Copper(II) and Cobalt(III) Complexes.
Acta chimica Slovenica (2016-12-23)
Ya-Li Sang, Xue-Song Lin, Wei-Dong Sun
ABSTRACT
A new trifluoromethylsulfonate salt of 2-[(3-chloropyridinium-2-yl)hydrazonomethyl]-6-methoxyphenol, (HL)CF3SO3, and its copper(II) and cobalt(III) complexes, [CuL(OH2)]CF3SO3 · 0.5H2O (1) and [CoL2]CF3SO3 · CH3OH (2), were prepared and characterized by physico-chemical methods and single crystal X-ray analysis. A trifluoromethylsulfonate anion is present in each of the compounds. The Cu atom in complex 1 is coordinated by the phenolate O, imino N and pyridine N atoms of L ligand, giving square planar geometry. The Co atom in complex 2 is coordinated by two phenolate O, two imino N and two pyridine N atoms from two L ligands, giving octahedral geometry. The three compounds were tested in vitro for their antibacterial activities.
Sigma-Aldrich
o-Vanillin, 99%