MilliporeSigma
  • Synthesis, biological characterization and evaluation of molecular mechanisms of novel copper complexes as anticancer agents.
Synthesis, biological characterization and evaluation of molecular mechanisms of novel copper complexes as anticancer agents.
Biochimica et biophysica acta (2016-10-25)
Ceyda Acilan, Buse Cevatemre, Zelal Adiguzel, Didem Karakas, Engin Ulukaya, Nádia Ribeiro, Isabel Correia, João Costa Pessoa
ABSTRACT
To overcome the hurdles of cisplatin, majorly its toxicity and resistance, there has been extensive search for alternative anti-cancer metal-based compounds. Here, three Cu(II)-complexes, Cu(Sal-Gly)(phen), Cu(Sal-Gly)(pheamine), Cu(Sal-Gly)(phepoxy) are characterized for their interaction with DNA, cytotoxicity and mechanism of action. The binding ability of the complexes to Calf-Thymus DNA was evaluated by competition fluorescence studies with thiazole-orange, UV-Vis and circular dichroism spectroscopic titrations. Cytotoxicity was evaluated by MTT analysis. The DNA damage was analyzed through cleavage of supercoiled DNA via agarose gel-electrophoresis, and 8-oxo-guanidine and ɣH2AX staining in cells. Apoptosis was detected via DNA condensation/fragmentation, mitochondrial membrane potential, Annexin V staining and caspase 3/7 activity. Formation of reactive oxygen species was determined by DCFDA- and GSSG/GSH-analysis. Binding constants to DNA were evaluated as 1.7×10 These Cu-complexes, namely Cu(Sal-Gly)(pheamine), may be considered promising anticancer agents with activity in cancer cells even with deficient p53 status.
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium/Nutrient Mixture F-12 Ham, With L-glutamine and trace elements, without HEPES and sodium bicarbonate, powder, suitable for cell culture
Sigma-Aldrich
Deoxyribonucleic acid sodium salt from calf thymus, suitable for substrate for DNase (Readily soluble)