MilliporeSigma
  • Anti-HSV-2 activity of Terminalia chebula Retz extract and its constituents, chebulagic and chebulinic acids.
Anti-HSV-2 activity of Terminalia chebula Retz extract and its constituents, chebulagic and chebulinic acids.
BMC complementary and alternative medicine (2017-02-16)
Ajay Kesharwani, Suja Kizhiyedath Polachira, Reshmi Nair, Aakanksha Agarwal, Nripendra Nath Mishra, Satish Kumar Gupta
ABSTRACT
Development of new and effective therapeutics for sexually transmitted herpes simplex virus-2 (HSV-2) infection is important from public health perspective. With an aim to identify natural products from medicinal plants, in the present study, the potential of Terminalia chebula Retz was investigated for its activity against HSV-2. Fruits of Terminalia chebula Retz were used to prepare 50% ethanolic extract. In addition, chebulagic acid and chebulinic acid both purified from T. chebula were also used. The extract as well as purified compounds were first used to determine their in vitro cytotoxicity on Vero cells by MTT assay. T. chebula extract, chebulagic acid, chebulinic acid along with acyclovir were subsequently assessed for direct anti-viral activity, and their ability to inhibit attachment and penetration of HSV-2 to the Vero cells. In addition, their anti-HSV-2 activity was also determined by in vitro post-infection plaque reduction assay. Cytotoxicity assay using Vero cells revealed CC The results presented herein suggest that T. chebula extract, chebulagic and chebulinic acids have higher direct antiviral activity against HSV-2 and efficacy to inhibit virus attachment and penetration to the host cells as compared to acyclovir. However, acyclovir is more potent to inhibit post-infection virus replication. Hence, T. chebula may be a useful candidate for developing alternative therapy for prevention of sexually transmitted HSV-2 infection. ᅟ.
Product Description
Supelco
Chebulagic acid, analytical standard
Pricing and availability is not currently available.