MilliporeSigma
  • Indoxyl sulfate - the uremic toxin linking hemostatic system disturbances with the prevalence of cardiovascular disease in patients with chronic kidney disease.
Indoxyl sulfate - the uremic toxin linking hemostatic system disturbances with the prevalence of cardiovascular disease in patients with chronic kidney disease.
BMC nephrology (2017-01-27)
Tomasz W Kamiński, Krystyna Pawlak, Małgorzata Karbowska, Michał Myśliwiec, Dariusz Pawlak
ABSTRACT
During chronic kidney disease progression, kidney-specific risk factors for cardiovascular disease come into play. The present study investigated the impact of indoxyl sulfate, dietary tryptophan-derived uremic toxin, accumulated in the blood of patients with chronic kidney disease on hemostatic parameters, markers of inflammation, oxidative stress and monocyte to macrophage transition. Fifty-one CKD patients not undergoing hemodialysis were enrolled in the study. Coagulation factors, fibrinolytic parameters, adhesion molecules, endothelial dysfunction markers, oxidative stress as well as inflammation markers were examined using immune-enzymatic method. Indoxyl sulfate levels were assessed using high-performance liquid chromatography. Biochemical parameters were determined by routine laboratory techniques using an automated analyzers. All assessed parameters were compared with controls and subjected to cross-sectional statistical analysis. Elevated concentrations of indoxyl sulfate, the vast majority of parameters affecting hemostasis, and markers of renal insufficiency conditions were observed. Part of hemostatic factors, namely tissue factor, von Willebrand factor, thrombomodulin, soluble urokinase-type plasminogen activator receptor, soluble intercellular adhesion molecule-1, soluble vascular cell adhesion protein were correlated with the fraction of indoxyl sulfate. A significant quantity of assessed parameters showed strong correlations with superoxide-dismutase, renal insufficiency rate, C-reactive protein, and neopterin. Levels of indoxyl sulfate were independently associated with markers of impaired endothelial function (thrombomodulin, adhesion molecules), oxidative stress (superoxide-dismutase) and monocytes activation determinant (neopterin), which indicate unconventional links between these systems and the role of indoxyl sulfate. Furthermore, parameters that correlated with the levels of indoxyl sulfate (von Willebrand factor, soluble urokinase-type plasminogen activator receptor, soluble intercellular adhesion molecule-1) were positively associated with the prevalence of cardiovascular disease in a CKD patients. The study demonstrated that in conditions of chronic exposure to uremic toxins, indoxyl sulfate seems to be one of the "missing links" between impaired renal function and prevalence of cardiovascular events, especially hemostatic disorders. The main functions of the action appear to be altered monocytes activation, intensified inflammatory process, and augmented oxidative stress by this uremic toxin.
Sigma-Aldrich
Rat CRP/C-Reactive Protein ELISA Kit, for serum, plasma and cell culture supernatant