MilliporeSigma
  • Derivatization of carbohydrates for chromatographic, electrophoretic and mass spectrometric structure analysis.
Derivatization of carbohydrates for chromatographic, electrophoretic and mass spectrometric structure analysis.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2003-07-26)
Fotini N Lamari, Reinhard Kuhn, Nikos K Karamanos
ABSTRACT
Carbohydrates, either alone or as constituents of glycoproteins, proteoglycans and glycolipids, are mediators of several cellular events and (patho)physiological processes. Progress in the "glycome" project is closely related to the analytical tools used to define carbohydrate structure and correlate structure with function. Chromatography, electrophoresis and mass spectrometry are the indispensable analytical tools of the on-going research. Carbohydrate derivatization is required for most of these analytical procedures. This review article gives an overview of derivatization methods of carbohydrates for their liquid chromatographic and electrophoretic separation, as well as the mass spectrometric characterization. Pre-column and on-capillary derivatization methods are presented with special emphasis on the derivatization of large carbohydrates.
Sigma-Aldrich
2-Aminopyridine, purum, ≥98.0% (NT)