MilliporeSigma
  • Small-window parametric imaging based on information entropy for ultrasound tissue characterization.
Small-window parametric imaging based on information entropy for ultrasound tissue characterization.
Scientific reports (2017-01-21)
Po-Hsiang Tsui, Chin-Kuo Chen, Wen-Hung Kuo, King-Jen Chang, Jui Fang, Hsiang-Yang Ma, Dean Chou
ABSTRACT
Constructing ultrasound statistical parametric images by using a sliding window is a widely adopted strategy for characterizing tissues. Deficiency in spatial resolution, the appearance of boundary artifacts, and the prerequisite data distribution limit the practicability of statistical parametric imaging. In this study, small-window entropy parametric imaging was proposed to overcome the above problems. Simulations and measurements of phantoms were executed to acquire backscattered radiofrequency (RF) signals, which were processed to explore the feasibility of small-window entropy imaging in detecting scatterer properties. To validate the ability of entropy imaging in tissue characterization, measurements of benign and malignant breast tumors were conducted (n = 63) to compare performances of conventional statistical parametric (based on Nakagami distribution) and entropy imaging by the receiver operating characteristic (ROC) curve analysis. The simulation and phantom results revealed that entropy images constructed using a small sliding window (side length = 1 pulse length) adequately describe changes in scatterer properties. The area under the ROC for using small-window entropy imaging to classify tumors was 0.89, which was higher than 0.79 obtained using statistical parametric imaging. In particular, boundary artifacts were largely suppressed in the proposed imaging technique. Entropy enables using a small window for implementing ultrasound parametric imaging.
Product Description
Supelco
Postcolumn Reactor Components, Glass Beads, size 75 μm, acid-washed
Pricing and availability is not currently available.