MilliporeSigma
  • Piperine regulates UCP1 through the AMPK pathway by generating intracellular lactate production in muscle cells.
Piperine regulates UCP1 through the AMPK pathway by generating intracellular lactate production in muscle cells.
Scientific reports (2017-01-25)
Nami Kim, Miso Nam, Mi Sun Kang, Jung Ok Lee, Yong Woo Lee, Geum-Sook Hwang, Hyeon Soo Kim
ABSTRACT
This study characterizes the human metabolic response to piperine, a curcumin extract, and the details of its underlying molecular mechanism. Using
Sigma-Aldrich
Guanosine 5′-diphosphate sodium salt, Type I, ≥96% (HPLC)
Sigma-Aldrich
Dorsomorphin, ≥98% (HPLC)