MilliporeSigma
Targeting SALL4 by entinostat in lung cancer.
Oncotarget (2016-10-06)
Kol Jia Yong, Ailing Li, Wen-Bin Ou, Clarice Kit Yee Hong, Wenxiu Zhao, Fei Wang, Hiro Tatetsu, Benedict Yan, Lihua Qi, Jonathan A Fletcher, Henry Yang, Ross Soo, Daniel G Tenen, Li Chai
ABSTRACT
The overall survival of lung cancer patients remains dismal despite the availability of targeted therapies. Oncofetal protein SALL4 is a novel cancer target. We herein report that SALL4 was aberrantly expressed in a subset of lung cancer patients with poor survival. SALL4 silencing by RNA interference or SALL4 peptide inhibitor treatment led to impaired lung cancer cell growth. Expression profiling of SALL4-knockdown cells demonstrated that both the EGFR and IGF1R signaling pathways were affected. Connectivity Map analysis revealed the HDAC inhibitor entinostat as a potential drug in treating SALL4-expressing cancers, and this was confirmed in 17 lung cancer cell lines. In summary, we report for the first time that entinostat can target SALL4-positive lung cancer. This lays the foundation for future clinical studies evaluating the therapeutic efficacy of entinostat in SALL4-positive lung cancer patients.
Sigma-Aldrich
MISSION® esiRNA, targeting human SALL4