MilliporeSigma
  • Supplementation With Cell-Penetrating Peptide-Conjugated Estrogen-Related Receptor β Improves the Formation of the Inner Cell Mass and the Development of Vitrified/Warmed Mouse Embryos.
Supplementation With Cell-Penetrating Peptide-Conjugated Estrogen-Related Receptor β Improves the Formation of the Inner Cell Mass and the Development of Vitrified/Warmed Mouse Embryos.
Reproductive sciences (Thousand Oaks, Calif.) (2016-04-14)
Ning Jie Yang, Dong-Won Seol, Junghyun Jo, Hyun Mee Jang, Sook-Young Yoon, Woo Sik Lee, Dong Ryul Lee
ABSTRACT
Estrogen-related receptor β (ESRRB), which is a member of the nuclear orphan receptor family, regulates the messenger RNA (mRNA) expression levels of the transcription factors, Oct4 and Nanog, in early embryos and germ cells, thereby maintaining the undifferentiated state and pluripotency of the relevant cells. The present study was designed to determine whether the upregulation of pluripotency-related genes by direct delivery of ESRRB protein may affect on the commitment into inner cell mass (ICM) or the development of vitrified/warmed mouse embryos. Recombinant cell-penetrating peptide (CPP) ESRRB protein was synthesized and then added into a culture medium for cryopreserved mouse embryos. Vitrified/warmed 8-cell embryos were cultured in KSOM with/without 2 μg/mL CPP-ESRRB for 48 hours and then analyzed or transferred to the uteri of foster mothers. The mRNA expression of Oct4 and Nanog was higher in CPP-ESRRB-treated blastocysts compared to the untreated controls. No difference was observed in embryonic development, but ICM:trophectoderm ratio was increased in the CPP-ESRRB-treated group compared to the untreated group, and after embryo transfer, a higher implantation rate was obtained in the CPP-ESRRB-treated group compared to the untreated group. This study shows for the first time that recombinant CPP-ESRRB can be easily integrated into vitrified/warmed mouse embryos and that it increases Oct4 expression (via a pluripotency-related gene pathway), ICM formation, and the further embryonic and full-term development of vitrified/warmed mouse embryos. This CPP-conjugated protein delivery system could therefore be a useful tool for improving assisted reproductive technology.
Sigma-Aldrich
TERGITOL, Type NP-10