MilliporeSigma
  • Omega-3 fatty acids, nutrient retention values, and sensory meat eating quality in cooked and raw Australian lamb.
Omega-3 fatty acids, nutrient retention values, and sensory meat eating quality in cooked and raw Australian lamb.
Meat science (2016-09-20)
Aaron Ross Flakemore, Bunmi Sherifat Malau-Aduli, Peter David Nichols, Aduli Enoch Othniel Malau-Aduli
ABSTRACT
This study evaluated omega-3 intramuscular fatty acids in the longissimus thoracis et lumborum of commercially prepared Australian lamb loin chops. Meats, denuded of external fats were cooked by means of conductive dry-heat using a fry grilling hot plate, to a core temperature of 70°C. An untrained consumer panel assessed meat appearance, aroma, tenderness, juiciness, taste and overall liking. Results showed no compositional alterations (P>0.05) to omega-3 fatty acids due to cooking treatment, whereas on absolute terms (mg/100g muscle) omega-3 fatty acids significantly (P<0.05) increased. The mean EPA+DHA content of the cooked meat at 32.8±2.3mg/100g muscle exceeded the minimum 30mg/100g per edible portion required for the defined Australian classification as 'source' long-chain (≥C20) omega-3 for cooked lamb. A 3.4% intramuscular fat content in the initial raw meat was sufficient to maintain acceptable overall sensory eating quality. Results endorse the application of this cooking method to enable delivery of health beneficial long-chain omega-3 fatty acids of commercially prepared Australian lamb loin chops to consumers without impediments to sensory eating properties.
Sigma-Aldrich
2-(Diisopropylamino)ethyl methacrylate, 97%, contains ~100 ppm monomethyl ether hydroquinone as inhibitor