MilliporeSigma
Efficacy of freeze-dried platelet-rich plasma in bone engineering.
Archives of oral biology (2016-10-25)
Yuya Nakatani, Hideki Agata, Yoshinori Sumita, Takamitsu Koga, Izumi Asahina
ABSTRACT
Platelet-rich plasma (PRP) is typically isolated and applied immediately after preparation, making it both a time- and labor-intensive addition to the operative procedure. Thus, it would be convenient if PRP could be preserved. We evaluated the efficacy of freeze-dried PRP (FD-PRP), as compared with freshly isolated PRP (f-PRP) for bone engineering. FD-PRP was prepared by lyophilization of f-PRP and was subsequently preserved at -20°C for one month. It was then rehydrated with an equal or 1/3 amount of distilled water (×1FD-PRP, ×3FD-PRP, respectively), and we assessed its gelation properties and the release of growth factors (PDGF-BB, TGF-β1, and VEGF). We also examined the bone forming ability with onlay-grafting on mice calvaria using β-TCP granules as a scaffold. FD-PRP showed comparable gelation as f-PRP. In terms of growth factor release,×1FD-PRP released identical concentrations of PDGF-BB and TGF-β1 to f-PRP, while ×3FD-PRP released approximately 3-fold concentrations when compared with f-PRP. In vivo, ×1FD-PRP promoted identical levels of the bone formation as f-PRP, and ×3FD-PRP induced more abundant bone formation. These results suggest that f-PRP can be stored without functional loss by freeze-drying and the concentration of PRP may improve its efficacy in bone engineering.
Sigma-Aldrich
Fibrinogen from bovine plasma, Type I-S, 65-85% protein (≥75% of protein is clottable)
Sigma-Aldrich
Ser-Phe-Leu-Leu-Arg-Asn-Pro-Asn-Asp-Lys-Tyr-Glu-Pro-Phe, ≥97% (HPLC)