MilliporeSigma
  • SILAC-based quantitative proteomic analysis of human lung cell response to copper oxide nanoparticles.
SILAC-based quantitative proteomic analysis of human lung cell response to copper oxide nanoparticles.
PloS one (2014-12-04)
Mariola J Edelmann, Leslie A Shack, Caitlin D Naske, Keisha B Walters, Bindu Nanduri
ABSTRACT
Copper (II) oxide (CuO) nanoparticles (NP) are widely used in industry and medicine. In our study we evaluated the response of BEAS-2B human lung cells to CuO NP, using Stable isotope labeling by amino acids in cell culture (SILAC)-based proteomics and phosphoproteomics. Pathway modeling of the protein differential expression showed that CuO NP affect proteins relevant in cellular function and maintenance, protein synthesis, cell death and survival, cell cycle and cell morphology. Some of the signaling pathways represented by BEAS-2B proteins responsive to the NP included mTOR signaling, protein ubiquitination pathway, actin cytoskeleton signaling and epithelial adherens junction signaling. Follow-up experiments showed that CuO NP altered actin cytoskeleton, protein phosphorylation and protein ubiquitination level.
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture