MilliporeSigma
  • Hippo/YAP signaling pathway is involved in osteosarcoma chemoresistance.
Hippo/YAP signaling pathway is involved in osteosarcoma chemoresistance.
Chinese journal of cancer (2016-05-22)
Dong-Yu Wang, Ya-Nan Wu, Jun-Qi Huang, Wei Wang, Meng Xu, Jin-Peng Jia, Gang Han, Bei-Bei Mao, Wen-Zhi Bi
ABSTRACT
Osteosarcoma is the most common bone malignancy in children and adolescents, and 20%-30% of the patients suffer from poor prognosis because of individual chemoresistance. The Hippo/yes-associated protein (YAP) signaling pathway has been shown to play a role in tumor chemoresistance, but no previous report has focused on its involvement in osteosarcoma chemoresistance. This study aimed to investigate the role of the Hippo/YAP signaling pathway in osteosarcoma chemoresistance and to determine potential treatment targets. Using the Cell Titer-Glo Luminescent cell viability assay and flow cytometry analysis, we determined the proliferation and chemosensitivity of YAP-overexpressing and YAP-knockdown osteosarcoma cells. In addition, using western blotting and the real-time polymerase chain reaction technique, we investigated the alteration of the Hippo/YAP signaling pathway in osteosarcoma cells treated with chemotherapeutic agents. Mammalian sterile 20-like kinase 1 (MST1) degradation was increased, and large tumor suppressor kinase 1/2 (LATS1/2) total protein levels were decreased by methotrexate and doxorubicin, which increased activation and nuclear translocation of YAP. Moreover, YAP increased the proliferation and chemoresistance of MG63 cells. The Hippo/YAP signaling pathway plays a role in osteosarcoma chemoresistance, and YAP is a potential target for reducing chemoresistance.
Sigma-Aldrich
Monoclonal Anti-TEAD4 antibody produced in mouse, clone 5H3, purified immunoglobulin, buffered aqueous solution