MilliporeSigma
Drep-2 is a novel synaptic protein important for learning and memory.
eLife (2014-11-14)
Till F M Andlauer, Sabrina Scholz-Kornehl, Rui Tian, Marieluise Kirchner, Husam A Babikir, Harald Depner, Bernhard Loll, Christine Quentin, Varun K Gupta, Matthew G Holt, Shubham Dipt, Michael Cressy, Markus C Wahl, André Fiala, Matthias Selbach, Martin Schwärzel, Stephan J Sigrist
ABSTRACT
CIDE-N domains mediate interactions between the DNase Dff40/CAD and its inhibitor Dff45/ICAD. In this study, we report that the CIDE-N protein Drep-2 is a novel synaptic protein important for learning and behavioral adaptation. Drep-2 was found at synapses throughout the Drosophila brain and was strongly enriched at mushroom body input synapses. It was required within Kenyon cells for normal olfactory short- and intermediate-term memory. Drep-2 colocalized with metabotropic glutamate receptors (mGluRs). Chronic pharmacological stimulation of mGluRs compensated for drep-2 learning deficits, and drep-2 and mGluR learning phenotypes behaved non-additively, suggesting that Drep 2 might be involved in effective mGluR signaling. In fact, Drosophila fragile X protein mutants, shown to benefit from attenuation of mGluR signaling, profited from the elimination of drep-2. Thus, Drep-2 is a novel regulatory synaptic factor, probably intersecting with metabotropic signaling and translational regulation.
Sigma-Aldrich
Anti-Rabbit-IgG - Atto 647N antibody produced in goat, 1 mg/mL IgG