MilliporeSigma
  • Microarray analysis on gene regulation by estrogen, progesterone and tamoxifen in human endometrial stromal cells.
Microarray analysis on gene regulation by estrogen, progesterone and tamoxifen in human endometrial stromal cells.
International journal of molecular sciences (2015-03-18)
Chun-E Ren, Xueqiong Zhu, Jinping Li, Christian Lyle, Sean Dowdy, Karl C Podratz, David Byck, Hai-Bin Chen, Shi-Wen Jiang
ABSTRACT
Epithelial stromal cells represent a major cellular component of human uterine endometrium that is subject to tight hormonal regulation. Through cell-cell contacts and/or paracrine mechanisms, stromal cells play a significant role in the malignant transformation of epithelial cells. We isolated stromal cells from normal human endometrium and investigated the morphological and transcriptional changes induced by estrogen, progesterone and tamoxifen. We demonstrated that stromal cells express appreciable levels of estrogen and progesterone receptors and undergo different morphological changes upon hormonal stimulation. Microarray analysis indicated that both estrogen and progesterone induced dramatic alterations in a variety of genes associated with cell structure, transcription, cell cycle, and signaling. However, divergent patterns of changes, and in some genes opposite effects, were observed for the two hormones. A large number of genes are identified as novel targets for hormonal regulation. These hormone-responsive genes may be involved in normal uterine function and the development of endometrial malignancies.
Sigma-Aldrich
Anti-Estrogen Receptor-β antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-Goat IgG (whole molecule)–Alkaline Phosphatase antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution