MilliporeSigma
  • Identification of cis- and trans-acting elements regulating calretinin expression in mesothelioma cells.
Identification of cis- and trans-acting elements regulating calretinin expression in mesothelioma cells.
Oncotarget (2016-02-06)
Jelena Kresoja-Rakic, Esra Kapaklikaya, Gabriela Ziltener, Damian Dalcher, Raffaella Santoro, Brock C Christensen, Kevin C Johnson, Beat Schwaller, Walter Weder, Rolf A Stahel, Emanuela Felley-Bosco
ABSTRACT
Calretinin (CALB2) is a diagnostic marker for epithelioid mesothelioma. It is also a prognostic marker since patients with tumors expressing high calretinin levels have better overall survival. Silencing of calretinin decreases viability of epithelioid mesothelioma cells. Our aim was to elucidate mechanisms regulating calretinin expression in mesothelioma. Analysis of calretinin transcript and protein suggested a control at the mRNA level. Treatment with 5-aza-2'-deoxycytidine and analysis of TCGA data indicated that promoter methylation is not likely to be involved. Therefore, we investigated CALB2 promoter by analyzing ~1kb of genomic sequence surrounding the transcription start site (TSS) + 1 using promoter reporter assay. Deletion analysis of CALB2 proximal promoter showed that sequence spanning the -161/+80bp region sustained transcriptional activity. Site-directed analysis identified important cis-regulatory elements within this -161/+80bp CALB2 promoter. EMSA and ChIP assays confirmed binding of NRF-1 and E2F2 to the CALB2 promoter and siRNA knockdown of NRF-1 led to decreased expression of calretinin. Cell synchronization experiment showed that calretinin expression was cell cycle regulated with a peak of expression at G1/S phase. This study provides the first insight in the regulation of CALB2 expression in mesothelioma cells.
Sigma-Aldrich
Anti-CALB2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-Cyclin A Antibody, Upstate®, from rabbit
Pricing and availability is not currently available.