MilliporeSigma
Atg7 cooperates with Pten loss to drive prostate cancer tumor growth.
Genes & development (2016-02-18)
Urmila Santanam, Whitney Banach-Petrosky, Cory Abate-Shen, Michael M Shen, Eileen White, Robert S DiPaola
ABSTRACT
Understanding new therapeutic paradigms for both castrate-sensitive and more aggressive castrate-resistant prostate cancer is essential to improve clinical outcomes. As a critically important cellular process, autophagy promotes stress tolerance by recycling intracellular components to sustain metabolism important for tumor survival. To assess the importance of autophagy in prostate cancer, we generated a new autochthonous genetically engineered mouse model (GEMM) with inducible prostate-specific deficiency in the Pten tumor suppressor and autophagy-related-7 (Atg7) genes. Atg7 deficiency produced an autophagy-deficient phenotype and delayed Pten-deficient prostate tumor progression in both castrate-naïve and castrate-resistant cancers. Atg7-deficient tumors display evidence of endoplasmic reticulum (ER) stress, suggesting that autophagy may promote prostate tumorigenesis through management of protein homeostasis. Taken together, these data support the importance of autophagy for both castrate-naïve and castrate-resistant growth in a newly developed GEMM, suggesting a new paradigm and model to study approaches to inhibit autophagy in combination with known and new therapies for advanced prostate cancer.
Sigma-Aldrich
Anti-Androgen Receptor antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Pricing and availability is not currently available.