MilliporeSigma
  • Expression of zinc finger E-box-binding homeobox factor 1 in epithelial ovarian cancer: A clinicopathological analysis of 238 patients.
Expression of zinc finger E-box-binding homeobox factor 1 in epithelial ovarian cancer: A clinicopathological analysis of 238 patients.
Molecular and clinical oncology (2016-02-13)
Xiufang Li, Ruixia Huang, Ruth Holm Li, Claes G Trope, Jahn M Nesland, Zhenhe Suo
ABSTRACT
A growing body of evidence indicates that aberrant activation of epithelial-to-mesenchymal transition (EMT) plays a key role in tumor cell invasion and metastasis. Zinc finger E-box-binding homeobox factor 1 (ZEB1), as a crucial mediator of EMT, contributes to the malignant progression of various epithelial tumors. To determine whether ZEB1 is involved in the progression of ovarian cancer, we immunohistochemically evaluated the expression of ZEB1 in 238 cases of epithelial ovarian cancer (EOC) and analyzed its associations with clinicopathological parameters. Positive expression of ZEB1 was observed in 32.8% (78/238) of EOCs and it was found to be significantly associated with advanced tumor stage (P=0.001). The survival analysis indicated that the expression of ZEB1 was associated with a poor 5-year progression-free survival (PFS) (P=0.021). A similar tendency was also observed between the expression of ZEB1 and 5-year overall survival, although it did not reach statistical significance (P=0.118). Moreover, the multivariate analysis demonstrated that ZEB1 expression was an independent risk factor for 5-year PFS in ovarian cancer. Taken together, our data provide evidence that ZEB1 may play a crucial role in promoting aggressive ovarian carcinoma progression. Therefore, ZEB1 may serve as an effectively predictive marker and a potential target for therapeutic intervention in EOC.
Sigma-Aldrich
Anti-ZEB1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution