MilliporeSigma
  • Alpha-synuclein inhibits aromatic amino acid decarboxylase activity in dopaminergic cells.
Alpha-synuclein inhibits aromatic amino acid decarboxylase activity in dopaminergic cells.
Journal of neurochemistry (2006-09-20)
Roya Tehranian, Susana E Montoya, Amber D Van Laar, Teresa G Hastings, Ruth G Perez
ABSTRACT
Alpha-synuclein is a presynaptic protein strongly implicated in Parkinson's disease (PD). Because dopamine neurons are invariably compromised during pathogenesis in PD, we have been exploring the functions of alpha-synuclein with particular relevance to dopaminergic neuronal cells. We previously discovered reduced tyrosine hydroxylase (TH) activity and minimal dopamine synthesis in stably-transfected MN9D cells overexpressing either wild-type or A53T mutant (alanine to threonine at amino acid 53) alpha-synuclein. TH, the rate-limiting enzyme in dopamine synthesis, converts tyrosine to l-dihydroxyphenylalanine (L-DOPA), which is then converted to dopamine by the enzyme, aromatic amino acid decarboxylase (AADC). We confirmed an interaction between alpha-synuclein and AADC in striatum. We then sought to determine whether wild-type or A53T mutant alpha-synuclein might have affected AADC activity in dopaminergic cells. Using HPLC with electrochemical detection, we measured dopamine and related catechols after L-DOPA treatments to bypass the TH step. We discovered that while alpha-synuclein did not reduce AADC protein levels, it significantly reduced AADC activity and phosphorylation in our cells. These novel findings further support a role for alpha-synuclein in dopamine homeostasis and may explain, at least in part, the selective vulnerability of dopamine neurons that occurs in PD.
Sigma-Aldrich
Anti-Tyrosine Hydroxylase Antibody, phosphoSer 40, Chemicon®, from rabbit
Pricing and availability is not currently available.