MilliporeSigma
  • A fluorescent probe for cysteine depalmitoylation reveals dynamic APT signaling.
A fluorescent probe for cysteine depalmitoylation reveals dynamic APT signaling.
Nature chemical biology (2016-12-20)
Rahul S Kathayat, Pablo D Elvira, Bryan C Dickinson
ABSTRACT
Hundreds of human proteins are modified by reversible palmitoylation of cysteine residues (S-palmitoylation), but the regulation of depalmitoylation is poorly understood. Here, we develop 'depalmitoylation probes' (DPPs), small-molecule fluorophores, to monitor the endogenous activity levels of 'erasers' of S-palmitoylation, acylprotein thioesterases (APTs). Live-cell analysis with DPPs reveals rapid growth-factor-mediated inhibition of the depalmitoylation activity of APTs, exposing a novel regulatory mechanism of dynamic lipid signaling.
Sigma-Aldrich
ML348, ≥98% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
ML349, ≥98% (HPLC)
Pricing and availability is not currently available.