MilliporeSigma
  • A neuronal lactate uptake inhibitor slows recovery of extracellular ion concentration changes in the hippocampal CA3 region by affecting energy metabolism.
A neuronal lactate uptake inhibitor slows recovery of extracellular ion concentration changes in the hippocampal CA3 region by affecting energy metabolism.
Journal of neurophysiology (2016-08-26)
Eskedar Ayele Angamo, Joerg Rösner, Agustin Liotta, Richard Kovács, Uwe Heinemann
ABSTRACT
Astrocyte-derived lactate supports pathologically enhanced neuronal metabolism, but its role under physiological conditions is still a matter of debate. Here, we determined the contribution of astrocytic neuronal lactate shuttle for maintenance of ion homeostasis and energy metabolism. We tested for the effects of α-cyano-4-hydroxycinnamic acid (4-CIN), which could interfere with energy metabolism by blocking monocarboxylate-transporter 2 (MCT2)-mediated neuronal lactate uptake, on evoked potentials, stimulus-induced changes in K
Sigma-Aldrich
CNQX, ≥98% (HPLC), solid
Supelco
Hydrogen ionophore II, Selectophore
Supelco
Calcium ionophore I, Selectophore, function tested, ≥99.0%