MilliporeSigma
  • EGF-Induced VEGF Exerts a PI3K-Dependent Positive Feedback on ERK and AKT through VEGFR2 in Hematological In Vitro Models.
EGF-Induced VEGF Exerts a PI3K-Dependent Positive Feedback on ERK and AKT through VEGFR2 in Hematological In Vitro Models.
PloS one (2016-11-03)
Lilian Saryeddine, Kazem Zibara, Nouhad Kassem, Bassam Badran, Nabil El-Zein
ABSTRACT
EGFR and VEGFR pathways play major roles in solid tumor growth and progression, however, little is known about these pathways in haematological tumors. This study investigated the crosstalk between EGFR and VEGFR2 signaling in two hematological in vitro models: THP1, a human monocytic leukemia, and Raji, a Burkitt's lymphoma, cell lines. Results showed that both cell lines express EGFR and VEGFR2 and responded to EGF stimulation by activating EGFR, triggering VEGF production and phosphorylating ERK, AKT, and p38 very early, with a peak of expression at 10-20min. Blocking EGFR using Tyrphostin resulted in inhibiting EGFR induced activation of ERK, AKT, and p38. In addition, EGF stimulation caused a significant and immediate increase, within 1min, in pVEGFR2 in both cell lines, which peaked at ~5-10 min after treatment. Selective inhibition of VEGFR2 by DMH4, anti-VEGFR2 antibody or siRNA diminished EGF-induced pAKT and pERK, indicating a positive feedback exerted by EGFR-induced VEGF. Similarly, the specific PI3K inhibitor LY294002, suppressed AKT and ERK phosphorylation showing that VEGF feedback is PI3K-dependent. On the other hand, phosphorylation of p38, initiated by EGFR and independent of VEGF feedback, was diminished using PLC inhibitor U73122. Moreover, measurement of intracellular [Ca2+] and ROS following VEGFR2 inhibition and EGF treatment proved that VEGFR2 is not implicated in EGF-induced Ca2+ release whereas it boosts EGF-induced ROS production. Furthermore, a significant decrease in pAKT, pERK and p-p38 was shown following the addition of the ROS inhibitor NAC. These results contribute to the understanding of the crosstalk between EGFR and VEGFR in haematological malignancies and their possible combined blockade in therapy.
Sigma-Aldrich
Fluo-3, suitable for fluorescence, ~70%
Sigma-Aldrich
MISSION® esiRNA, targeting human KDR