MilliporeSigma
  • Balancing potency, metabolic stability and permeability in pyrrolopyrimidine-based EGFR inhibitors.
Balancing potency, metabolic stability and permeability in pyrrolopyrimidine-based EGFR inhibitors.
European journal of medicinal chemistry (2016-09-12)
Jin Han, Silje Henriksen, Kristin G Nørsett, Eirik Sundby, Bård Helge Hoff
ABSTRACT
The present study describes our continuous effort to develop epidermal growth factor receptor (EGFR) inhibitors based on the 6-aryl-pyrrolo[2,3-d]pyrimidin-4-amine scaffold. The activity-ADME space has been evaluated by synthesizing 43 new structures, including four variations of the 4-amino group and 34 different substitution patterns in the 6-aryl moiety. Most of the new pyrrolopyrimidines were highly active, with twelve analogues possessing lower IC
Sigma-Aldrich
(S)-(+)-2-Phenylglycinol, 98%