MilliporeSigma
  • Stanniocalcin 1 Enhances Carbon Flux from Glucose to Lipids in White Retroperitoneal Adipose Tissue in the Fed Rat.
Stanniocalcin 1 Enhances Carbon Flux from Glucose to Lipids in White Retroperitoneal Adipose Tissue in the Fed Rat.
Lipids (2016-10-25)
Aline G Cozer, Márcia Trapp, Claudia Vieira Marques, Tiago L Martins, Jorge Felipe A Model, Vanessa Schein, Luiz Carlos Kucharski, Roselis S M Da Silva
ABSTRACT
The present work assesses in vitro the role of human Stanniocalcin 1 (hSTC-1) in glucose metabolism in white retroperitoneal adipose tissue (WRAT) from fed rat. In the fed state, hSTC1 increases the incorporation of
Sigma-Aldrich
ADP/ATP Ratio Assay Kit, sufficient for 100 tests (bioluminescent)