MilliporeSigma
  • Requirement of JNK for stress-induced activation of the cytochrome c-mediated death pathway.
Requirement of JNK for stress-induced activation of the cytochrome c-mediated death pathway.
Science (New York, N.Y.) (2000-05-08)
C Tournier, P Hess, D D Yang, J Xu, T K Turner, A Nimnual, D Bar-Sagi, S N Jones, R A Flavell, R J Davis
ABSTRACT
The c-Jun NH2-terminal kinase (JNK) is activated when cells are exposed to ultraviolet (UV) radiation. However, the functional consequence of JNK activation in UV-irradiated cells has not been established. It is shown here that JNK is required for UV-induced apoptosis in primary murine embryonic fibroblasts. Fibroblasts with simultaneous targeted disruptions of all the functional Jnk genes were protected against UV-stimulated apoptosis. The absence of JNK caused a defect in the mitochondrial death signaling pathway, including the failure to release cytochrome c. These data indicate that mitochondria are influenced by proapoptotic signal transduction through the JNK pathway.
Sigma-Aldrich
JNK1, active, GST tagged from mouse, PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥90% (SDS-PAGE), buffered aqueous glycerol solution
Pricing and availability is not currently available.