MilliporeSigma
  • Pilot study of the pharmacokinetics of betel nut and betel quid biomarkers in saliva, urine, and hair of betel consumers.
Pilot study of the pharmacokinetics of betel nut and betel quid biomarkers in saliva, urine, and hair of betel consumers.
Drug testing and analysis (2015-12-02)
Adrian A Franke, Xingnan Li, Jennifer F Lai
ABSTRACT
Approximately 600 million people worldwide practise the carcinogenic habit of betel nut/quid chewing. Carcinogenic N-nitroso compounds have been identified in saliva or urine of betel chewers and the betel alkaloid arecoline in hair from habitual betel quid chewers. However, the pharmacokinetic parameters of these compounds have been little explored. Assessment of betel use by biomarkers is urgently needed to evaluate the effectiveness of cessation programmes aimed at reducing betel consumption to decrease the burden of cancers in regions of high betel consumption. In the search for biomarkers of betel consumption, we measured by liquid chromatography-mass spectrometry (LC-MS) the appearance and disappearance of betel alkaloids (characteristic for betel nuts), N-nitroso compounds, and chavibetol (characteristic for Piper Betle leaves) in saliva (n=4), hair (n=2), and urine (n=1) of occasional betel nut/quid chewers. The betel alkaloids arecoline, guvacoline, guvacine, and arecaidine were detected in saliva of all four participants and peaked within the first 2 h post-chewing before returning to baseline levels after 8 h. Salivary chavibetol was detected in participants consuming Piper Betle leaves in their quid and peaked ~1 h post-chewing. Urinary arecoline, guvacoline, and arecaidine excretion paralleled saliva almost exactly while chavibetol glucuronide excretion paralleled salivary chavibetol. No betel nut related compounds were detected in the tested hair samples using various extraction methods. From these preliminary results, we conclude that betel exposure can only be followed on a short-term basis (≤8 h post-chewing) using the applied biomarkers from urine and saliva while the feasibility of using hair has yet to be validated. Copyright © 2015 John Wiley & Sons, Ltd.
Product Description
Supelco
N-Nitrosoanatabine, analytical standard
Pricing and availability is not currently available.