MilliporeSigma
  • Development & automation of a novel [(18)F]F prosthetic group, 2-[(18)F]-fluoro-3-pyridinecarboxaldehyde, and its application to an amino(oxy)-functionalised Aβ peptide.
Development & automation of a novel [(18)F]F prosthetic group, 2-[(18)F]-fluoro-3-pyridinecarboxaldehyde, and its application to an amino(oxy)-functionalised Aβ peptide.
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine (2016-08-16)
Olivia Morris, J Gregory, M Kadirvel, Fiona Henderson, A Blykers, Adam McMahon, Mark Taylor, David Allsop, Stuart Allan, J Grigg, Herve Boutin, Christian Prenant
ABSTRACT
2-[(18)F]-Fluoro-3-pyridinecarboxaldehyde ([(18)F]FPCA) is a novel, water-soluble prosthetic group. It's radiochemistry has been developed and fully-automated for application in chemoselective radiolabelling of amino(oxy)-derivatised RI-OR2-TAT peptide, (Aoa-k)-RI-OR2-TAT, using a GE TRACERlab FX-FN. RI-OR2-TAT is a brain-penetrant, retro-inverso peptide that binds to amyloid species associated with Alzheimer's Disease. Radiolabelled (Aoa-k)-RI-OR2-TAT was reproducibly synthesised and the product of the reaction with FPCA has been fully characterised. In-vivo biodistribution of [(18)F]RI-OR2-TAT has been measured in Wistar rats.