MilliporeSigma
Hydrogen Evolution Catalyzed by Cu
ACS applied materials & interfaces (2016-09-10)
Faruk Ozel, Emre Aslan, Adem Sarilmaz, Imren Hatay Patir
ABSTRACT
The present study reports, for the first time, both a facile synthesis for ternary Cu
Sigma-Aldrich
Tungsten(IV) chloride, 95%
Pricing and availability is not currently available.