MilliporeSigma
  • Impact of Environmental Chemicals on the Transcriptome of Primary Human Hepatocytes: Potential for Health Effects.
Impact of Environmental Chemicals on the Transcriptome of Primary Human Hepatocytes: Potential for Health Effects.
Journal of biochemical and molecular toxicology (2016-04-20)
Robert D Mitchell, Anirudh Dhammi, Andrew Wallace, Ernest Hodgson, R Michael Roe
ABSTRACT
New paradigms for human health risk assessment of environmental chemicals emphasize the use of molecular methods and human-derived cell lines. In this study, we examined the effects of the insect repellent DEET (N,N-diethyl-m-toluamide) and the phenylpyrazole insecticide fipronil (fluocyanobenpyrazole) on transcript levels in primary human hepatocytes. These chemicals were tested individually and as a mixture. RNA-Seq showed that 100 μM DEET significantly increased transcript levels (α = 0.05) for 108 genes and lowered transcript levels for 64 genes and fipronil at 10 μM increased the levels of 2246 transcripts and decreased the levels for 1428 transcripts. Fipronil was 21-times more effective than DEET in eliciting changes, even though the treatment concentration was 10-fold lower for fipronil versus DEET. The mixture of DEET and fipronil produced a more than additive effect (levels increased for 3017 transcripts and decreased for 2087 transcripts). The transcripts affected for all chemical treatments were classified by GO analysis and mapped to chromosomes. The overall treatment responses, specific pathways, and individual transcripts affected were discussed at different levels of fold-change. Changes found in transcript levels in response to treatments will require further research to understand their importance in overall cellular, organ, and organismic function.
Product Description
SAFC
Williams′ Medium E, With sodium bicarbonate, without L-glutamine and phenol red, liquid, sterile-filtered, suitable for cell culture
Pricing and availability is not currently available.