MilliporeSigma
  • Versatile and efficient synthesis of omega-functionalized asymmetric disulfides via sulfenyl bromide adducts.
Versatile and efficient synthesis of omega-functionalized asymmetric disulfides via sulfenyl bromide adducts.
Langmuir : the ACS journal of surfaces and colloids (2007-01-26)
Janusz Kowalczyk, Piotr Barski, Dariusz Witt, Bartosz A Grzybowski
ABSTRACT
Various types of asymmetric disulfides can be synthesized under mild conditions and in excellent yields by a method involving dialkoxylthiophosphoranesulfenyl halide precursors. This straightforward, rapid procedure is used to prepare a series of disulfides bearing neutral, acidic, and basic terminal groups as well as groups commonly used in biospecific self-assembled monolayers.
Sigma-Aldrich
Triethylene glycol mono-11-mercaptoundecyl ether, 95%
Pricing and availability is not currently available.