MilliporeSigma
  • Genetically determined dosage of follicle-stimulating hormone (FSH) affects male reproductive parameters.
Genetically determined dosage of follicle-stimulating hormone (FSH) affects male reproductive parameters.
The Journal of clinical endocrinology and metabolism (2011-07-08)
Marina Grigorova, Margus Punab, Birutė Zilaitienė, Juris Erenpreiss, Kristo Ausmees, Valentinas Matuleviĉius, Igor Tsarev, Niels Jørgensen, Maris Laan
ABSTRACT
The detailed role of FSH in contributing to male testicular function and fertility has been debated. We have previously identified the association between the T-allele of the FSHB promoter polymorphism (rs10835638; G/T, -211 bp from the mRNA start) and significantly reduced male serum FSH. In the current study, the T-allele carriers of the FSHB -211 G/T single nucleotide polymorphism represented a natural model for documenting downstream phenotypic consequences of insufficient FSH action. We genotyped rs10835638 in the population-based Baltic cohort of young men (n = 1054; GG carriers, n = 796; GT carriers, n = 244; TT carriers, n = 14) recruited by Andrology Centres in Tartu, Estonia; Riga, Latvia; and Kaunas, Lithuania. Marker-trait association testing was performed using linear regression (additive, recessive models) adjusted by age, body mass index, smoking, and recruitment center. Serum hormones directly correlated with the T-allele dosage of rs10835638 included FSH (additive model, P = 1.11 × 10(-6); T-allele effect, -0.41 IU/liter), inhibin-B (P = 2.16 × 10(-3); T-allele effect, -14.67 pg/ml), and total testosterone (P = 9.30 × 10(-3); T-allele effect, -1.46 nmol/liter). Parameters altered only among TT homozygotes were reduced testicular volume (recessive model, P = 1.19 × 10(-4); TT genotype effect, -9.47 ml) and increased serum LH (P = 2.25 × 10(-2); TT genotype effect, 1.07 IU/liter). The carrier status of rs10835638 alternative genotypes did not affect sperm motility and morphology, calculated free testosterone, serum SHBG, and estradiol concentrations. We showed for the first time that genetically determined low FSH may have wider downstream effects on the male reproductive system, including impaired testes development, altered testicular hormone levels (inhibin-B, total testosterone, LH), and affected male reproductive potential.