MilliporeSigma
  • A role for mitogen-activated protein kinase in mediating activation of the glycoprotein hormone alpha-subunit promoter by gonadotropin-releasing hormone.
A role for mitogen-activated protein kinase in mediating activation of the glycoprotein hormone alpha-subunit promoter by gonadotropin-releasing hormone.
Molecular and cellular biology (1995-07-01)
M S Roberson, A Misra-Press, M E Laurance, P J Stork, R A Maurer
ABSTRACT
Gonadotropin-releasing hormone (GnRH) interacts with a G protein-coupled receptor and increases the transcription of the glycoprotein hormone alpha-subunit gene. We have explored the possibility that mitogen-activated protein kinase (MAPK) plays a role in mediating GnRH effects on transcription. Activation of the MAPK cascade by an expression vector for a constitutively active form of the Raf-1 kinase led to stimulation of the alpha-subunit promoter in a concentration-dependent manner. GnRH treatment was found to increase the phosphorylation of tyrosine residues of MAPK and to increase MAPK activity, as determined by an immune complex kinase assay. A reporter gene assay using the MAPK-responsive, carboxy-terminal domain of the Elk1 transcription factor was also consistent with GnRH-induced activation of MAPK. Interference with the MAPK pathway by expression vectors for kinase-defective MAPKs or vectors encoding MAPK phosphatases reduced the transcription-stimulating effects of GnRH. The DNA sequences which are required for responses to GnRH include an Ets factor-binding site. An expression vector for a dominant negative form of Ets-2 was able to reduce GnRH effects on expression of the alpha-subunit gene. These findings provide evidence that GnRH treatment leads to activation of the MAPK cascade in gonadotropes and that activation of MAPK contributes to stimulation of the alpha-subunit promoter. It is likely that an Ets factor serves as a downstream transcriptional effector of MAPK in this system.