MilliporeSigma
  • Novel 8-Hydroxyquinoline Derivatives as Multitarget Compounds for the Treatment of Alzheimer's Disease.
Novel 8-Hydroxyquinoline Derivatives as Multitarget Compounds for the Treatment of Alzheimer's Disease.
ChemMedChem (2016-02-18)
Federica Prati, Christian Bergamini, Romana Fato, Ondrej Soukup, Jan Korabecny, Vincenza Andrisano, Manuela Bartolini, Maria Laura Bolognesi
ABSTRACT
We discovered a small series of hit compounds that show multitargeting activities against key targets in Alzheimer's disease (AD). The compounds were designed by combining the structural features of the anti-AD drug donepezil with clioquinol, which is able to chelate redox-active metals, thus decreasing metal-driven oxidative phenomena and β-amyloid (Aβ)-mediated neurotoxicity. The majority of the new hybrid compounds selectively target human butyrylcholinesterase at micromolar concentrations and effectively inhibit Aβ self-aggregation. In addition, compounds 5-chloro-7-((4-(2-methoxybenzyl)piperazin-1-yl)methyl)-8-hydroxyquinoline (1 b), 7-((4-(2-methoxybenzyl)piperazin-1-yl)methyl)-8-hydroxyquinoline (2 b), and 7-(((1-benzylpiperidin-4-yl)amino)methyl)-5-chloro-8-hydroxyquinoline (3 a) are able to chelate copper(II) and zinc(II) and exert antioxidant activity in vitro. Importantly, in the case of 2 b, the multitarget profile is accompanied by high predicted blood-brain barrier permeability, low cytotoxicity in T67 cells, and acceptable toxicity in HUVEC primary cells.
Sigma-Aldrich
4-Amino-1-benzylpiperidine, 98%