MilliporeSigma
  • A Cullin3-KLHL20 Ubiquitin ligase-dependent pathway targets PML to potentiate HIF-1 signaling and prostate cancer progression.
A Cullin3-KLHL20 Ubiquitin ligase-dependent pathway targets PML to potentiate HIF-1 signaling and prostate cancer progression.
Cancer cell (2011-08-16)
Wei-Chien Yuan, Yu-Ru Lee, Shiu-Feng Huang, Yu-Min Lin, Tzu-Yin Chen, Hsiang-Ching Chung, Chin-Hsien Tsai, Hsin-Yi Chen, Cheng-Ta Chiang, Chun-Kai Lai, Li-Ting Lu, Chun-Hau Chen, De-Leung Gu, Yeong-Shiau Pu, Yuh-Shan Jou, Kun Ping Lu, Pei-Wen Hsiao, Hsiu-Ming Shih, Ruey-Hwa Chen
ABSTRACT
Tumor hypoxia is associated with disease progression and treatment failure, but the hypoxia signaling mechanism is not fully understood. Here, we show that KLHL20, a Cullin3 (Cul3) substrate adaptor induced by HIF-1, coordinates with the actions of CDK1/2 and Pin1 to mediate hypoxia-induced PML proteasomal degradation. Furthermore, this PML destruction pathway participates in a feedback mechanism to maximize HIF-1α induction, thereby potentiating multiple tumor hypoxia responses, including metabolic reprogramming, epithelial-mesenchymal transition, migration, tumor growth, angiogenesis, and chemoresistance. In human prostate cancer, overexpression of HIF-1α, KLHL20, and Pin1 correlates with PML down-regulation, and hyperactivation of the PML destruction pathway is associated with disease progression. Our study indicates that the KLHL20-mediated PML degradation and HIF-1α autoregulation play key roles in tumor progression.