MilliporeSigma
  • Highly luminescent S, N co-doped graphene quantum dots with broad visible absorption bands for visible light photocatalysts.
Highly luminescent S, N co-doped graphene quantum dots with broad visible absorption bands for visible light photocatalysts.
Nanoscale (2013-10-24)
Dan Qu, Min Zheng, Peng Du, Yue Zhou, Ligong Zhang, Di Li, Huaqiao Tan, Zhao Zhao, Zhigang Xie, Zaicheng Sun
ABSTRACT
A facile hydrothermal synthesis route to N and S, N co-doped graphene quantum dots (GQDs) was developed by using citric acid as the C source and urea or thiourea as N and S sources. Both N and S, N doped GQDs showed high quantum yield (78% and 71%), excitation independent under excitation of 340-400 nm and single exponential decay under UV excitation. A broad absorption band in the visible region appeared in S, N co-doped GQDs due to doping with sulfur, which alters the surface state of GQDs. However, S, N co-doped GQDs show different color emission under excitation of 420-520 nm due to their absorption in the visible region. The excellent photocatalytic performance of the S, N co-doped GQD/TiO2 composites was demonstrated by degradation of rhodamine B under visible light. The apparent rate of S, N:GQD/TiO2 is 3 and 10 times higher than that of N:GQD/TiO2 and P25 TiO2 under visible light irradiation, respectively.
Sigma-Aldrich
Nitrogen/Sulfur co-doped graphene, avg. no. of layers, 1 ‑ 5