MilliporeSigma
  • Oligodeoxynucleotide inhibition of Toll-like receptors 3, 7, 8, and 9 suppresses cytokine production in a human rheumatoid arthritis model.
Oligodeoxynucleotide inhibition of Toll-like receptors 3, 7, 8, and 9 suppresses cytokine production in a human rheumatoid arthritis model.
European journal of immunology (2015-11-26)
Sandra Sacre, Alexandra Lo, Bernard Gregory, Matthew Stephens, Giselle Chamberlain, Philip Stott, Fionula Brennan
ABSTRACT
Toll-like receptors (TLRs) are innate immune receptors that respond to both exogenous and endogenous stimuli and are suggested to contribute to the perpetuation of chronic inflammation associated with rheumatoid arthritis (RA). In particular, the endosomal TLRs 3, 7, 8, and 9 have more recently been postulated to be of importance in RA pathogenesis. In this study, pan inhibition of the endosomal TLRs by a phosphorothioate-modified inhibitory oligodeoxynucleotide (ODN) is demonstrated in primary human B cells, macrophages, and RA fibroblasts. Inhibition of TLR8 was of particular interest as TLR8 has been associated with RA pathogenesis in both human and murine arthritis models. ODN1411 competitively inhibited TLR8 signaling and was observed to directly bind to a purified TLR8 ectodomain, suggesting inhibition was through a direct interaction with the receptor. Addition of ODN1411 to human RA synovial membrane cultures significantly inhibited spontaneous cytokine production from these cultures, suggesting a potential role for one or more of the endosomal TLRs in inflammatory cytokine production in RA and the potential for inhibitory ODNs as novel therapies.
Sigma-Aldrich
Insect GeneJuice® Transfection Reagent, Proprietary liposome transfection reagent optimized for maximal transfection efficiency of Sf9 insect cells for baculovirus protein expression.