MilliporeSigma
  • Aminomethylations via cross-coupling of potassium organotrifluoroborates with aryl bromides.
Aminomethylations via cross-coupling of potassium organotrifluoroborates with aryl bromides.
Organic letters (2007-03-21)
Gary A Molander, Deidre L Sandrock
ABSTRACT
[reaction: see text] The Suzuki-Miyaura cross-coupling reaction of N,N-dialkylaminomethyltrifluoroborates with aryl halides allows the construction of an aminomethyl aryl linkage through a disconnection based on dissonant reactivity patterns. A variety of these aminomethyltrifluoroborate substrates were prepared in good to excellent yields and then shown to cross-couple with equal facility to both electron-rich and electron-poor aryl halides as well as to a variety of heteroaromatic bromides.
Sigma-Aldrich
5-Bromo-2-furaldehyde, 97%