MilliporeSigma
  • Microwave assisted Leimgruber-Batcho reaction for the preparation of indoles, azaindoles and pyrroylquinolines.
Microwave assisted Leimgruber-Batcho reaction for the preparation of indoles, azaindoles and pyrroylquinolines.
Organic & biomolecular chemistry (2004-01-23)
Jason Siu, Ian R Baxendale, Steven V Ley
ABSTRACT
The development of enhanced conditions for Lewis acid catalysed Leimgruber-Batcho indole synthesis using microwave acceleration is described. This approach has permitted the preparation of a variety of heteroaromatic enamine intermediates in good yield and high purities. Subsequent catalytic hydrogenation reactions, under various conditions including the use of a solid-phase encapsulated catalyst, furnish the corresponding indole derivatives in good yields.
Sigma-Aldrich
1H-Benzo[g]indole, 97%
Pricing and availability is not currently available.