MilliporeSigma
  • The Effect of a Selective Inhibitor of Phosphodiesterase-9 on Oxidative Stress, Inflammation and Cytotoxicity in Neutrophils from Patients with Sickle Cell Anaemia.
The Effect of a Selective Inhibitor of Phosphodiesterase-9 on Oxidative Stress, Inflammation and Cytotoxicity in Neutrophils from Patients with Sickle Cell Anaemia.
Basic & clinical pharmacology & toxicology (2015-09-09)
Maritza Cavalcante Barbosa, Talyta Ellen de Jesus dos Santos, Thayna Nogueira dos Santos, Alano Martins Pedrosa, Darcielle Bruna Dias Elias, Luzia Kalyne Almeida Moreira Leal, Amanda de Araújo Lopes, Greyce Luri Sasahara, Romélia Pinheiro Gonçalves Lemes
ABSTRACT
The aim of the study was to investigate the possible anti-inflammatory and antioxidant effects of BAY 73-6691 on neutrophils from SCA patients. This study included 35 patients with a molecular diagnosis of SCA, whose neutrophils were isolated and treated with BAY 73-6691 at the concentrations 100, 10, 1.0 and 0.1 μg/mL. LDH release and MTT assays were performed to verify cell viability. To evaluate oxidative stress, the following parameters were determined by spectrophotometric assays: NO and malondialdehyde (MDA) levels and activity of catalase, superoxide dismutase (SOD) and glutathione peroxidase (GPx). As inflammatory markers, myeloperoxidase (MPO) levels were evaluated by colorimetric assay and TNF-α by enzyme immunoassay. The results showed that neutrophils from SCA patients not treated with hydroxyurea (HU) had significantly lower NO levels and catalase and SOD activity, as well as significantly higher MDA, MPO and TNF-α levels when compared with neutrophils from SCA patients treated with HU and neutrophils from control group. Treatment of SCA neutrophils with BAY 73-6691 resulted in 94%, 200% and 168% increase in NOx levels, SOD and catalase activity, respectively. In addition, there was a reduction of approximately 46% and 45% in TNF-α and MPO levels, respectively. In SCAHU neutrophils, there was a 30% and 44% increase in NOx levels and SOD activity, respectively, and a 28% and 37% decrease in TNF-α and MPO levels, respectively. However, these effects were observed at cytotoxic doses only. The results of this study are original and demonstrate that inhibition of phosphodiesterase-9 in neutrophils from SCA patients with BAY 73-6691 was able to increase the NO bioavailability and attenuate oxidative stress and inflammation in neutrophils from patients not treated with HU.
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
BAY 73-6691, ≥98% (HPLC), powder