MilliporeSigma
  • DLC1 activation requires lipid interaction through a polybasic region preceding the RhoGAP domain.
DLC1 activation requires lipid interaction through a polybasic region preceding the RhoGAP domain.
Molecular biology of the cell (2009-08-28)
Patrik Erlmann, Simone Schmid, Florian A Horenkamp, Matthias Geyer, Thomas G Pomorski, Monilola A Olayioye
ABSTRACT
Deleted in Liver Cancer 1 (DLC1) is a GTPase-activating protein (GAP) with specificity for RhoA, RhoB, and RhoC that is frequently deleted in various tumor types. By inactivating these small GTPases, DLC1 controls actin cytoskeletal remodeling and biological processes such as cell migration and proliferation. Here we provide evidence that DLC1 binds to phosphatidylinositol-4,5-bisphosphate (PI(4,5)P(2)) through a previously unrecognized polybasic region (PBR) adjacent to its RhoGAP domain. Importantly, PI(4,5)P(2)-containing membranes are shown to stimulate DLC1 GAP activity in vitro. In living cells, a DLC1 mutant lacking an intact PBR inactivated Rho signaling less efficiently and was severely compromised in suppressing cell spreading, directed migration, and proliferation. We therefore propose that PI(4,5)P(2) is an important cofactor in DLC1 regulation in vivo and that the PBR is essential for the cellular functions of the protein.
Product Description
Roche
ABTS Buffer, solution, pkg of 125 mL