MilliporeSigma
  • Functional xenobiotic metabolism and efflux transporters in trout hepatocyte spheroid cultures.
Functional xenobiotic metabolism and efflux transporters in trout hepatocyte spheroid cultures.
Toxicology research (2015-04-22)
Chibuzor Uchea, Stewart F Owen, J Kevin Chipman
ABSTRACT
Prediction of xenobiotic fate in fish is important for the regulatory assessment of chemicals under current legislation. Trout hepatocyte spheroids are a promising