MilliporeSigma
  • Highly sensitive PCR assay for routine diagnosis of African swine fever virus in clinical samples.
Highly sensitive PCR assay for routine diagnosis of African swine fever virus in clinical samples.
Journal of clinical microbiology (2003-09-06)
M Agüero, J Fernández, L Romero, C Sánchez Mascaraque, M Arias, J M Sánchez-Vizcaíno
ABSTRACT
This work provides a novel, highly sensitive, hot start PCR method for rapid and specific detection of African swine fever virus (ASFV) that can be used as a routine diagnostic test for ASFV in surveillance, control, and eradication programs. A confirmatory test of the specificity of this method based on restriction endonuclease analysis was also developed.
Product Description
Roche
DNA Molecular Weight Marker VI, DIG-labeled, solution, pkg of 500 μL (10 μg/ml)