MilliporeSigma
  • Polymerase chain reaction combined with restriction enzyme analysis for detection and differentiation of fowl adenoviruses.
Polymerase chain reaction combined with restriction enzyme analysis for detection and differentiation of fowl adenoviruses.
Avian pathology : journal of the W.V.P.A (2001-12-01)
G Meulemans, M Boschmans, T P Berg, M Decaesstecker
ABSTRACT
A polymerase chain reaction combined with restriction enzyme analysis was developed for detection and differentiation of all 12 fowl adenovirus (FAdV) serotypes representing the five fowl adenovirus (A to E) species. For primer design, the published sequences of the hexon proteins of FAdV1, FAdV8 and FAdV9 were aligned and conserved regions in the two pedestal regions adjacent to the L1 loop region were determined. A primer pair (hexon A/hexon B) was constructed and was shown to amplify approximately 900 bp of the hexon gene including the L1 loop region. An amplification product was detected using supernatant of infected cell cultures from all FAdV1 to FAdV12 reference strains used in our study. The sequence and the restriction patterns of the hexon A/B fragments of the 12 FAdV strains were determined and compared. The successive use of four different endonucleases allowed the complete differentiation of the reference FAdV strains. Twenty-six fowl adenoviruses isolated during our routine virological diagnosis activities could all be amplified using hexon A/hexon B primers. Restriction analysis results showed that 8/26 adenovirus strains contained two different FAdV types. FAdV4, FAdV12, FAdV1, FAdV5 and FAdV6 were the most frequently isolated.
Product Description
Roche
DNA Molecular Weight Marker VI, pkg of 50 μg (in 200 μl), solution
Pricing and availability is not currently available.