MilliporeSigma
  • Genetic engineering of Synechocystis PCC6803 for the photoautotrophic production of the sweetener erythritol.
Genetic engineering of Synechocystis PCC6803 for the photoautotrophic production of the sweetener erythritol.
Microbial cell factories (2016-04-10)
Aniek D van der Woude, Ruth Perez Gallego, Angie Vreugdenhil, Vinod Puthan Veetil, Tania Chroumpi, Klaas J Hellingwerf
ABSTRACT
Erythritol is a polyol that is used in the food and beverage industry. Due to its non-caloric and non-cariogenic properties, the popularity of this sweetener is increasing. Large scale production of erythritol is currently based on conversion of glucose by selected fungi. In this study, we describe a biotechnological process to produce erythritol from light and CO2, using engineered Synechocystis sp. PCC6803. By functionally expressing codon-optimized genes encoding the erythrose-4-phosphate phosphatase TM1254 and the erythrose reductase Gcy1p, or GLD1, this cyanobacterium can directly convert the Calvin cycle intermediate erythrose-4-phosphate into erythritol via a two-step process and release the polyol sugar in the extracellular medium. Further modifications targeted enzyme expression and pathway intermediates. After several optimization steps, the best strain, SEP024, produced up to 2.1 mM (256 mg/l) erythritol, excreted in the medium.
Product Description
Roche
Anti-His6, from mouse IgG1
Pricing and availability is not currently available.