MilliporeSigma
  • Supramolecular luminescence from oligofluorenol-based supramolecular polymer semiconductors.
Supramolecular luminescence from oligofluorenol-based supramolecular polymer semiconductors.
International journal of molecular sciences (2013-11-16)
Guang-Wei Zhang, Long Wang, Ling-Hai Xie, Jin-Yi Lin, Wei Huang
ABSTRACT
Supramolecular luminescence stems from non-covalent exciton behaviors of active π-segments in supramolecular entities or aggregates via intermolecular forces. Herein, a π-conjugated oligofluorenol, containing self-complementary double hydrogen bonds, was synthesized using Suzuki coupling as a supramolecular semiconductor. Terfluorenol-based random supramolecular polymers were confirmed via concentration-dependent nuclear magnetic resonance (NMR) and dynamic light scattering (DLS). The photoluminescent spectra of the TFOH-1 solution exhibit a green emission band (g-band) at approximately ~520 nm with reversible features, as confirmed through titration experiments. Supramolecular luminescence of TFOH-1 thin films serves as robust evidence for the aggregates of g-band. Our results suggest that the presence of polyfluorene ketone defects is a sufficient condition, rather than a sufficient-necessary condition for the g-band. Supramolecular electroluminescence will push organic devices into the fields of supramolecular optoelectronics, spintronics, and mechatronics.
Sigma-Aldrich
2,2′-Bithiophene, 99%
Sigma-Aldrich
2,7-Dibromo-9-fluorenone, 96%