MilliporeSigma
  • The clinical significance of DC-SIGN and DC-SIGNR, which are novel markers expressed in human colon cancer.
The clinical significance of DC-SIGN and DC-SIGNR, which are novel markers expressed in human colon cancer.
PloS one (2014-12-17)
Yanmei Jiang, Changfu Zhang, Kai Chen, Zhe Chen, Zhigang Sun, Zhuqing Zhang, Dongbing Ding, Shuangyi Ren, Yunfei Zuo
ABSTRACT
Colon cancer has always been diagnosed at a late stage, which is associated with poor prognosis. The currently used serum tumor markers CEA and CA19-9 display low sensitivity and specificity and may not have diagnostic value in early stage colon cancer. Thus, there is an urgent need to identify novel serum biomarkers for use in the early detection of colon cancer. In this study, the expression of DC-SIGN and DC-SIGNR in serum was detected by enzyme-linked immunosorbent assay (ELISA). DC-SIGN and DC-SIGNR expression was detected in cancer tissues by immunohistochemistry (IHC). The level of sDC-SIGN was lower in patients than in the healthy controls, while the level of sDC-SIGNR in patients was higher than in the healthy controls. Both sDC-SIGN and sDC-SIGNR had diagnostic significances for cancer patients, and the combined diagnosis of these two markers was higher than both of them alone. Furthermore, there were significant differences between both sDC-SIGN and sDC-SIGNR in stage I/II patients and the healthy controls. Moreover, high sDC-SIGN level was accompanied with the long survival time. Additionally, DC-SIGNR was negative in the cancer foci and matched normal colon tissues but was weakly positive between the cancer foci. DC-SIGN staining was faint in matched normal colon tissues, strong in the tumor stroma and the invasive margin of colon cancer tissues, and negatively correlated with the sDC-SIGN level in serum from the same patient. Interestingly, the percent survival of patients with a DC-SIGN mean density of>0.001219 (the upper 95% confidence interval of matched normal colon tissues) was higher than for all other patients. DC-SIGN and DC-SIGNR are blood-based molecular markers that can potentially be used for the diagnosis of early stage patients. Moreover, expression of DC-SIGN in serum and cancer tissues may affect the survival time for colon cancer patients.
Sigma-Aldrich
MISSION® esiRNA, targeting human CD209