MilliporeSigma
Copper-catalyzed intermolecular hydroamination of alkenes.
Organic letters (2006-07-28)
Jason G Taylor, Neil Whittall, King Kuok Mimi Hii
ABSTRACT
[reaction: see text] Regioselective additions of arylsulfonamides to vinylarenes, norbornene, and cyclohexadiene were achieved using a copper-diphosphine catayst under mild reaction conditions. These processes appear to be ligand-accelerated.
Sigma-Aldrich
rac-BINAP, 97%